siRNA Details
| Gene Symbol: CLEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099431.1 | NP_001092901.1 | 5 | 681 | ||
| NM_016509.3 | NP_057593.3 | 6 | 780 | |||
| XM_005253382.4 | XP_005253439.1 | 8 | 1152 | |||
| XM_011520685.2 | XP_011518987.1 | 7 | 1061 | |||
| XM_017019395.1 | XP_016874884.1 | 7 | 1136 | |||
| XM_017019396.1 | XP_016874885.1 | 5 | 488 | |||
| GenBank mRNA | AF124841.1 | - | 6 | 637 | ||
| AY358599.1 | - | 6 | 761 | |||
| BC029554.1 | - | 7 | 752 | 5269252 | ||

CLEC1B
Genomic Map