siRNA Details
| Gene Symbol: IER5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016545.4 | NP_057629.2 | 1 | 1234 | ||
| GenBank mRNA | AF178984.1 | - | 1 | 975 | ||
| AF258581.1 | - | 1 | 1022 | |||
| AJ251089.1 | - | 1 | 1064 | |||
| AK314830.1 | - | 1 | 1036 | |||
| BC000128.2 | - | 1 | 1024 | 3048356 | ||
| BC108253.1 | - | 1 | 1020 | 6025403 | ||

IER5
Genomic Map