siRNA Details
| Gene Symbol: BFAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016561.2 | NP_057645.1 | 3 | 657 | ||
| GenBank mRNA | AF173003.1 | - | 3 | 529 | ||
| AK291114.1 | - | 3 | 465 | |||
| AK297067.1 | - | 3 | 465 | |||
| AK313890.1 | - | 2 | 465 | |||
| BC003054.1 | - | 3 | 488 | 3537783 | ||
| BI912683.1 | - | 3 | 657 | |||

BFAR
Genomic Map