siRNA Details
| Gene Symbol: SLC15A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016582.2 | NP_057666.1 | 2 | 843 | ||
| NR_027391.1 | 2 | 843 | ||||
| XM_011545095.2 | XP_011543397.1 | 2 | 1138 | |||
| GenBank mRNA | AB020598.1 | - | 2 | 843 | ||
| AF135600.1 | - | 2 | 736 | |||
| BC037974.1 | - | 2 | 703 | 5213632 | ||

SLC15A3
Genomic Map