siRNA Details
| Gene Symbol: FAM53C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135647.1 | NP_001129119.1 | 4 | 1374 | ||
| NM_016605.2 | NP_057689.1 | 4 | 1349 | |||
| GenBank mRNA | AF251040.1 | - | 4 | 1085 | ||
| AK027412.1 | - | 2 | 240 | |||
| AK310378.1 | - | 4 | 527 | |||
| AK315567.1 | - | 4 | 1098 | |||
| BC000259.1 | - | 4 | 1091 | 3357346 | ||
| BC052993.1 | - | 4 | 1314 | 5164799 | ||

FAM53C
Genomic Map