siRNA Details
| Gene Symbol: FAM53C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135647.1 | NP_001129119.1 | 3 | 580 | ||
| NM_016605.2 | NP_057689.1 | 3 | 555 | |||
| GenBank mRNA | AF251040.1 | - | 3 | 291 | ||
| AK026729.1 | - | 2 | 700 | |||
| AK310378.1 | - | 3 | 304 | |||
| AK315567.1 | - | 3 | 304 | |||
| BC000259.1 | - | 3 | 297 | 3357346 | ||
| BC052993.1 | - | 3 | 520 | 5164799 | ||
| DA216192.1 | - | 2 | 325 | |||

FAM53C
Genomic Map