siRNA Details
| Gene Symbol: NME8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016616.4 | NP_057700.3 | 10 | 974 | ||
| GenBank mRNA | AF202051.2 | - | 9 | 703 | ||
| AF305596.1 | - | 9 | 672 | |||
| AK313599.1 | - | 9 | 714 | |||
| BC036816.1 | - | 10 | 974 | 5745079 | ||
| GQ472221.1 | - | 10 | 974 | |||

NME8
Genomic Map