siRNA Details
| Gene Symbol: CDH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016279.3 | NP_057363.3 | 3 | 601 | ||
| GenBank mRNA | AB035302.1 | - | 3 | 586 | ||
| AK307779.1 | - | 3 | 601 | |||
| AK308697.1 | - | 3 | 605 | |||
| BC107594.1 | - | 3 | 563 | 40011942 | ||
| BC113745.1 | - | 3 | 563 | 40011941 | ||

CDH9
Genomic Map