siRNA Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016523.2 | NP_057607.1 | 3 | 325 | ||
| XM_017019414.1 | XP_016874903.1 | 3 | 444 | |||
| XM_017019415.1 | XP_016874904.1 | 3 | 516 | |||
| GenBank mRNA | AF175206.1 | - | 3 | 325 | ||
| AJ305370.1 | - | 3 | 308 | |||
| BC096734.1 | - | 3 | 285 | |||
| BC098166.1 | - | 3 | 285 | 40015535 | ||
| BC098257.1 | - | 3 | 285 | 40015534 | ||
| BC098354.1 | - | 3 | 285 | 40015531 | ||

KLRF1
Genomic Map