siRNA Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291822.1 | NP_001278751.1 | 3 | 340 | ||
| NM_016523.2 | NP_057607.1 | 4 | 490 | |||
| GenBank mRNA | AF175206.1 | - | 4 | 490 | ||
| AF175207.1 | - | 3 | 340 | |||
| AJ305370.1 | - | 4 | 473 | |||
| BC098354.1 | - | 4 | 450 | 40015531 | ||

KLRF1
Genomic Map