siRNA Details
Gene Symbol: TMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF161448.1 | - | 3 | 397 | ||
AK126594.1 | - | 4 | 2308 | |||
AL712164.1 | - | 4 | 425 | |||
AY794023.1 | - | 4 | 432 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF161448.1 | - | 3 | 397 | ||
AK126594.1 | - | 4 | 2308 | |||
AL712164.1 | - | 4 | 425 | |||
AY794023.1 | - | 4 | 432 |