siRNA Details
| Gene Symbol: RWDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007464.2 | NP_001007465.1 | 6 | 772 | ||
| NM_015952.3 | NP_057036.2 | 5 | 658 | |||
| NM_016104.3 | NP_057188.2 | 7 | 900 | |||
| GenBank mRNA | AF092134.1 | - | 5 | 615 | ||
| AF132958.1 | - | 5 | 522 | |||
| AK290727.1 | - | 5 | 658 | |||
| AK301089.1 | - | 6 | 635 | |||
| BC015802.1 | - | 5 | 514 | 3909348 | ||
| CF128570.1 | - | 5 | 605 | |||

RWDD1
Genomic Map