siRNA Details
| Gene Symbol: RWDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007464.2 | NP_001007465.1 | 8 | 952 | ||
| NM_015952.3 | NP_057036.2 | 7 | 838 | |||
| NM_016104.3 | NP_057188.2 | 9 | 1080 | |||
| GenBank mRNA | AF092134.1 | - | 7 | 795 | ||
| AF132958.1 | - | 7 | 702 | |||
| AK290727.1 | - | 7 | 838 | |||
| AK301089.1 | - | 8 | 815 | |||
| BC015802.1 | - | 7 | 694 | 3909348 | ||

RWDD1
Genomic Map