siRNA Details
| Gene Symbol: AIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286587.1 | NP_001273516.1 | 5 | 450 | ||
| NM_016108.3 | NP_057192.2 | 4 | 507 | |||
| XM_011535883.2 | XP_011534185.1 | 4 | 488 | |||
| XM_011535884.2 | XP_011534186.1 | 4 | 490 | |||
| XM_017010927.1 | XP_016866416.1 | 4 | 506 | |||
| GenBank mRNA | AF151861.1 | - | 5 | 453 | ||
| AF153605.1 | - | 4 | 450 | |||
| AJ420480.1 | - | 2 | 123 | |||
| AK001347.1 | - | 4 | 448 | |||
| BC025278.1 | - | 4 | 438 | 4859216 | ||
| BF725860.1 | - | 3 | 340 | |||
| BX538067.1 | - | 3 | 210 | |||

AIG1
Genomic Map