siRNA Details
| Gene Symbol: AIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286587.1 | NP_001273516.1 | 4 | 373 | ||
| NM_001286588.1 | NP_001273517.1 | 3 | 430 | |||
| NM_001286589.1 | NP_001273518.1 | 3 | 430 | |||
| NM_016108.3 | NP_057192.2 | 3 | 430 | |||
| XM_011535881.2 | XP_011534183.1 | 3 | 412 | |||
| XM_011535882.2 | XP_011534184.1 | 3 | 411 | |||
| XM_011535883.2 | XP_011534185.1 | 3 | 411 | |||
| XM_011535884.2 | XP_011534186.1 | 3 | 413 | |||
| XM_011535885.2 | XP_011534187.1 | 3 | 411 | |||
| XM_017010927.1 | XP_016866416.1 | 3 | 429 | |||
| XM_017010928.1 | XP_016866417.1 | 3 | 411 | |||
| GenBank mRNA | AF151861.1 | - | 4 | 376 | ||
| AF153605.1 | - | 3 | 373 | |||
| AJ420480.1 | - | 1 | 46 | |||
| AK001347.1 | - | 3 | 371 | |||
| AK222475.1 | - | 3 | 392 | |||
| AK298533.1 | - | 3 | 430 | |||
| BC025278.1 | - | 3 | 361 | 4859216 | ||
| BF725860.1 | - | 2 | 263 | |||
| BX538067.1 | - | 2 | 133 | |||
| BX640703.1 | - | 3 | 372 | |||
| DB154031.1 | - | 3 | 352 | |||

AIG1
Genomic Map