siRNA Details
| Gene Symbol: WDR83OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016145.3 | NP_057229.1 | 2, 3 | 399 | ||
| XM_017026864.1 | XP_016882353.1 | 3, 4 | 1393 | |||
| GenBank mRNA | AF059620.1 | - | 2, 3 | 167 | ||
| AF078861.1 | - | 2, 3 | 384 | |||
| AF151898.1 | - | 2, 3 | 383 | |||
| AK091030.1 | - | 2, 3 | 2104 | |||
| AK311944.1 | - | 2, 3 | 163 | |||
| BC001192.1 | - | 2, 3 | 147 | |||
| BC042919.1 | - | 2, 3 | 168 | 5932109 | ||
| BC061912.1 | - | 2, 3 | 168 | 4444565 | ||

WDR83OS
Genomic Map