siRNA Details
| Gene Symbol: SNX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016224.4 | NP_057308.1 | 6 | 851 | ||
| XM_005267015.2 | XP_005267072.1 | 8 | 929 | |||
| XM_011535886.2 | XP_011534188.1 | 3 | 390 | |||
| GenBank mRNA | AB209736.1 | - | 2 | 182 | ||
| AF001629.1 | - | 6 | 656 | |||
| AF076957.1 | - | 1 | 71 | |||
| AF121859.1 | - | 6 | 611 | |||
| AF131214.1 | - | 6 | 630 | |||
| AF172847.1 | - | 6 | 731 | |||
| AK026637.1 | - | 3 | 396 | |||
| AK127345.1 | - | 3 | 706 | |||
| AK314360.1 | - | 6 | 685 | |||
| BC005022.1 | - | 6 | 758 | 3832234 | ||

SNX9
Genomic Map