siRNA Details
| Gene Symbol: MAGEC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016249.3 | NP_057333.1 | 3 | 571 | ||
| GenBank mRNA | AF116194.1 | - | 3 | 554 | ||
| AF151378.1 | - | 3 | 552 | |||
| AF196482.1 | - | 3 | 552 | |||
| AK313591.1 | - | 3 | 564 | |||
| BC005891.2 | - | 1 | 302 | 3050294 | ||
| BC013318.1 | - | 3 | 575 | 4133159 | ||
| CR457168.1 | - | 1 | 223 | |||

MAGEC2
Genomic Map