siRNA Details
| Gene Symbol: FAM8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016255.2 | NP_057339.1 | 3 | 1063 | ||
| GenBank mRNA | AF097027.1 | - | 3 | 995 | ||
| AK296393.1 | - | 4 | 377 | |||
| AK307681.1 | - | 3 | 555 | |||
| AK312814.1 | - | 3 | 998 | |||
| BC047881.1 | - | 3 | 987 | 5742372 | ||

FAM8A1
Genomic Map