siRNA Details
| Gene Symbol: VGLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016267.3 | NP_057351.1 | 4 | 817 | ||
| GenBank mRNA | AF137387.1 | - | 4 | 713 | ||
| AK309912.1 | - | 4 | 754 | |||
| BC000045.1 | - | 4 | 817 | 3504527 | ||
| BC003362.1 | - | 4 | 715 | 2900293 | ||

VGLL1
Genomic Map