siRNA Details
Gene Symbol: DCDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195610.1 | NP_001182539.1 | 10 | 1248 | ||
NM_016356.4 | NP_057440.2 | 9 | 1341 | |||
GenBank mRNA | AB032980.2 | - | 9 | 1039 | ||
AF181721.1 | - | 9 | 1341 | |||
AK027036.1 | - | 9 | 1157 | |||
AL133043.1 | - | 2 | 320 | |||
BC050704.1 | - | 10 | 1248 | 5758845 |
