siRNA Details
| Gene Symbol: TEN1-CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037709.1 | 8 | 2405 | |||
| GenBank mRNA | AK097104.1 | - | 8 | 2362 | ||
| BX647274.1 | - | 8 | 2391 | |||
| Gene Symbol: CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258.2 | NP_001249.1 | 6 | 645 | ||
| GenBank mRNA | X66357.1 | - | 5 | 657 | ||

TEN1-CDK3
Genomic Map