siRNA Details
| Gene Symbol: SPOUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016390.3 | NP_057474.2 | 6, 7 | 548 | ||
| XM_017014804.1 | XP_016870293.1 | 7, 8 | 815 | |||
| GenBank mRNA | AL110193.1 | - | 6, 7 | 548 | ||
| BC010579.1 | - | 6, 7 | 510 | 4213299 | ||
| BC021273.1 | - | 6, 7 | 521 | 4843384 | ||
| BC033677.1 | - | 6, 7 | 516 | 5176852 | ||
| BC039590.1 | - | 6, 7 | 533 | 5757547 | ||
| BC046133.1 | - | 6, 7 | 513 | 5725479 | ||
| BC063644.1 | - | 6, 7 | 516 | 4539701 | ||

SPOUT1
Genomic Map