siRNA Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016470.7 | NP_057554.4 | 4 | 664 | ||
| XM_011528851.1 | XP_011527153.1 | 5 | 1139 | |||
| XM_011528853.2 | XP_011527155.1 | 4 | 708 | |||
| XM_011528854.2 | XP_011527156.1 | 3 | 447 | |||
| XM_017027873.1 | XP_016883362.1 | 5 | 1204 | |||
| GenBank mRNA | AF151041.1 | - | 4 | 547 | ||
| AF161517.1 | - | 4 | 547 | |||
| AF217514.1 | - | 4 | 530 | |||
| AK000476.1 | - | 4 | 568 | |||
| AK315155.1 | - | 4 | 529 | |||
| AL133000.1 | - | 4 | 549 | |||
| BC064973.1 | - | 4 | 514 | 6151501 | ||

OSER1
Genomic Map