siRNA Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016470.7 | NP_057554.4 | 4 | 952 | ||
| XM_011528851.1 | XP_011527153.1 | 5 | 1427 | |||
| XM_011528853.2 | XP_011527155.1 | 4 | 996 | |||
| XM_011528854.2 | XP_011527156.1 | 3 | 735 | |||
| XM_017027873.1 | XP_016883362.1 | 5 | 1492 | |||
| GenBank mRNA | AF151041.1 | - | 4 | 836 | ||
| AF161517.1 | - | 4 | 835 | |||
| AF217514.1 | - | 4 | 819 | |||
| AK000476.1 | - | 4 | 857 | |||
| AK315155.1 | - | 4 | 817 | |||
| AL133000.1 | - | 4 | 837 | |||
| BC064973.1 | - | 4 | 802 | 6151501 | ||

OSER1
Genomic Map