siRNA Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016481.4 | NP_057565.3 | 5 | 1264 | ||
| XM_005252048.4 | XP_005252105.1 | 6 | 1324 | |||
| XM_005252049.4 | XP_005252106.1 | 4 | 1046 | |||
| XM_005252050.4 | XP_005252107.1 | 5 | 1150 | |||
| XM_011518777.1 | XP_011517079.1 | 6 | 1303 | |||
| GenBank mRNA | AK000213.1 | - | 5 | 1264 | ||
| AK023069.1 | - | 2 | 2633 | |||
| AY189280.1 | - | 7 | 3317 | |||
| BC002863.2 | - | 5 | 1244 | 3944148 | ||
| X86033.1 | - | 2 | 536 | |||

TRMO
Genomic Map