siRNA Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016481.4 | NP_057565.3 | 3, 4 | 494 | ||
| XM_005252047.3 | XP_005252104.1 | 3, 4 | 451 | |||
| XM_005252049.4 | XP_005252106.1 | 2, 3 | 276 | |||
| XM_011518776.2 | XP_011517078.1 | 3, 4 | 451 | |||
| GenBank mRNA | AF151053.1 | - | 3, 4 | 440 | ||
| AK000213.1 | - | 3, 4 | 494 | |||
| BC002863.2 | - | 3, 4 | 474 | 3944148 | ||

TRMO
Genomic Map