siRNA Details
| Gene Symbol: ZCCHC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282566.1 | NP_001269495.1 | 3 | 306 | ||
| NM_001282567.1 | NP_001269496.1 | 3 | 168 | |||
| NM_001282568.1 | NP_001269497.1 | 4 | 427 | |||
| NM_001282569.1 | NP_001269498.1 | 5 | 359 | |||
| NM_001282570.1 | NP_001269499.1 | 3 | 168 | |||
| NM_001282572.1 | NP_001269501.1 | 4 | 289 | |||
| NM_001282573.1 | NP_001269502.1 | 5 | 359 | |||
| NM_001282574.1 | NP_001269503.1 | 4 | 289 | |||
| NM_016505.3 | NP_057589.2 | 4 | 289 | |||
| XM_006710681.3 | XP_006710744.1 | 5 | 433 | |||
| XM_011541567.2 | XP_011539869.1 | 4 | 305 | |||
| GenBank mRNA | AF151077.1 | - | 4 | 218 | ||
| AF247661.2 | - | 3 | 158 | |||
| AK001929.1 | - | 4 | 239 | |||
| AK022506.1 | - | 4 | 276 | |||
| AK024049.1 | - | 4 | 265 | |||
| AK302248.1 | - | 5 | 309 | |||
| AK302343.1 | - | 3 | 114 | |||
| AK307478.1 | - | 3 | 145 | |||
| BC000685.2 | - | 4 | 327 | |||
| BC007446.2 | - | 4 | 225 | 3009823 | ||
| BC050609.1 | - | 4 | 229 | 6527025 | ||
| BF309574.1 | - | 4 | 240 | |||
| CN423139.1 | - | 4 | 226 | |||
| DA737244.1 | - | 5 | 309 | |||
| DC366453.1 | - | 4 | 427 | |||

ZCCHC17
Genomic Map