siRNA Details
| Gene Symbol: RAB6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016577.3 | NP_057661.3 | 2, 3 | 517 | ||
| XM_011512893.2 | XP_011511195.1 | 2, 3 | 192 | |||
| GenBank mRNA | AF091031.1 | - | 1, 2 | 64 | ||
| AF166492.1 | - | 2, 3 | 473 | |||
| AF498940.1 | - | 2, 3 | 136 | |||
| AK295451.1 | - | 3, 4 | 254 | |||
| AK312378.1 | - | 2, 3 | 437 | |||
| BC002510.2 | - | 2, 3 | 486 | 3050592 | ||
| BC078662.1 | - | 2, 3 | 265 | 6450986 | ||
| BT007263.1 | - | 2, 3 | 136 | |||

RAB6B
Genomic Map