siRNA Details
| Gene Symbol: ESF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276380.1 | NP_001263309.1 | 2 | 708 | ||
| NM_016649.3 | NP_057733.2 | 2 | 719 | |||
| XM_017027874.1 | XP_016883363.1 | 2 | 2171 | |||
| GenBank mRNA | AF068285.1 | - | 1 | 76 | ||
| AK000375.1 | - | 2 | 720 | |||
| AK022369.1 | - | 2 | 670 | |||
| AK074299.1 | - | 1 | 205 | |||
| BC020952.1 | - | 1 | 338 | |||
| BC030513.1 | - | 1 | 669 | |||
| BC039706.1 | - | 1 | 660 | |||
| BC046107.1 | - | 1 | 658 | |||

ESF1
Genomic Map