siRNA Details
| Gene Symbol: ESF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276380.1 | NP_001263309.1 | 2 | 140 | ||
| NM_016649.3 | NP_057733.2 | 2 | 151 | |||
| XM_017027874.1 | XP_016883363.1 | 2 | 1603 | |||
| GenBank mRNA | AK000375.1 | - | 2 | 151 | ||
| AK022369.1 | - | 2 | 102 | |||
| BC030513.1 | - | 1 | 101 | |||
| BC039706.1 | - | 1 | 92 | |||
| BC046107.1 | - | 1 | 90 | |||
| BC062214.1 | - | 2 | 130 | |||
| BC104663.1 | - | 1 | 100 | |||
| DA472223.1 | - | 2 | 140 | |||

ESF1
Genomic Map