siRNA Details
| Gene Symbol: CUTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014433.2 | NP_001014433.1 | 1 | 173 | ||
| NM_001014837.1 | NP_001014837.1 | 1 | 173 | |||
| NM_001014838.1 | NP_001014838.1 | 1 | 173 | |||
| NM_001014840.1 | NP_001014840.1 | 1 | 173 | |||
| NM_015921.2 | NP_057005.1 | 1 | 173 | |||
| GenBank mRNA | CN363642.1 | - | 1 | 203 | ||

CUTA
Genomic Map