siRNA Details
| Gene Symbol: LIPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204830.1 | NP_001191759.1 | 3 | 1487 | ||
| NM_015929.3 | NP_057013.1 | 3 | 1289 | |||
| NM_145197.2 | NP_660198.1 | 4 | 1417 | |||
| NM_145198.2 | NP_660199.1 | 4 | 1405 | |||
| NM_145199.2 | NP_660200.1 | 2 | 1163 | |||
| NR_037935.1 | 4 | 1550 | ||||
| NR_037936.1 | 3 | 1226 | ||||
| GenBank mRNA | AB017566.1 | - | 3 | 1289 | ||
| BC007001.1 | - | 3 | 1209 | 3961227 | ||
| BC009772.1 | - | 3 | 1230 | 4091817 | ||
| BC070145.1 | - | 3 | 1267 | 4671058 | ||

LIPT1
Genomic Map