siRNA Details
| Gene Symbol: TAF9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015975.4 | NP_057059.2 | 6 | 596 | ||
| GenBank mRNA | AF220509.1 | - | 6 | 547 | ||
| AK312579.1 | - | 6 | 555 | |||
| BC009566.1 | - | 6 | 547 | 3996687 | ||
| BC010350.1 | - | 6 | 561 | 4276335 | ||
| BC071649.1 | - | 6 | 533 | 5246715 | ||
| BC146952.1 | - | 6 | 577 | 9056880 | ||
| BC146960.1 | - | 6 | 577 | 9056888 | ||
| BI544376.1 | - | 6 | 625 | |||

TAF9B
Genomic Map