siRNA Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015983.3 | NP_057067.1 | 5 | 315 | ||
| XM_006715734.3 | XP_006715797.1 | 5 | 315 | |||
| XM_011515420.2 | XP_011513722.1 | 6 | 408 | |||
| XM_011515422.2 | XP_011513724.1 | 4 | 252 | |||
| XM_011515423.2 | XP_011513725.1 | 2 | 99 | |||
| XM_017012292.1 | XP_016867781.1 | 6 | 408 | |||
| XM_017012293.1 | XP_016867782.1 | 6 | 408 | |||
| XM_017012294.1 | XP_016867783.1 | 6 | 408 | |||
| XM_017012295.1 | XP_016867784.1 | 3 | 162 | |||
| GenBank mRNA | AF125044.1 | - | 5 | 354 | ||
| AF125045.1 | - | 4 | 201 | |||
| AK001446.1 | - | 5 | 303 | |||
| BC004104.2 | - | 5 | 307 | 3835609 | ||
| BG499454.1 | - | 5 | 330 | |||

UBE2D4
Genomic Map