siRNA Details
| Gene Symbol: DYNC2LI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193464.1 | NP_001180393.1 | 4 | 301 | ||
| NM_015522.3 | NP_056337.1 | 4 | 301 | |||
| NM_016008.3 | NP_057092.2 | 4 | 301 | |||
| XM_005264364.4 | XP_005264421.1 | 4 | 278 | |||
| XM_005264365.4 | XP_005264422.1 | 4 | 278 | |||
| GenBank mRNA | AF151818.1 | - | 4 | 262 | ||
| AK074206.1 | - | 4 | 291 | |||
| AK223432.1 | - | 4 | 301 | |||
| AL050006.1 | - | 3 | 154 | |||
| AY083346.1 | - | 4 | 261 | |||
| BC006969.1 | - | 4 | 260 | 3828551 | ||
| BC016324.1 | - | 4 | 295 | 4093010 | ||
| BC040558.1 | - | 4 | 276 | |||
| BC058823.1 | - | 4 | 268 | 4093493 | ||
| BG716986.1 | - | 4 | 306 | |||
| BX493698.1 | - | 4 | 304 | |||

DYNC2LI1
Genomic Map