siRNA Details
| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016047.3 | NP_057131.1 | 2 | 264 | ||
| GenBank mRNA | AF151868.1 | - | 2 | 260 | ||
| AF161523.1 | - | 2 | 158 | |||
| AF184213.1 | - | 2 | 260 | |||
| AF401310.1 | - | 2 | 49 | |||
| AK000621.1 | - | 2 | 164 | |||
| AK311908.1 | - | 2 | 197 | |||
| BC015463.1 | - | 2 | 264 | 3914184 | ||

SF3B6
Genomic Map