siRNA Details
| Gene Symbol: NUDT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001248011.1 | NP_001234940.1 | 4 | 781 | ||
| NM_024047.4 | NP_076952.1 | 5 | 877 | |||
| NM_198038.2 | NP_932155.1 | 5 | 519 | |||
| XM_011532032.2 | XP_011530334.1 | 5 | 3056 | |||
| GenBank mRNA | AF265341.1 | - | 5 | 518 | ||
| AF273028.1 | - | 5 | 841 | |||
| AK027295.1 | - | 5 | 867 | |||
| AK074845.1 | - | 5 | 519 | |||
| AK075408.1 | - | 5 | 797 | |||
| AK308028.1 | - | 4 | 548 | |||
| AY026252.1 | - | 5 | 872 | |||
| AY359123.1 | - | 5 | 571 | |||
| BC000542.2 | - | 5 | 637 | 3163944 | ||
| BI552154.1 | - | 518 | ||||
| BM544951.1 | - | 5 | 887 | |||

NUDT9
Genomic Map