siRNA Details
| Gene Symbol: TM6SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001524.2 | NP_001001524.2 | 3 | 279 | ||
| GenBank mRNA | AF255923.1 | - | 3 | 228 | ||
| AK097070.1 | - | 3 | 279 | |||
| BC120986.1 | - | 3 | 253 | |||
| BC120987.1 | - | 3 | 253 | |||
| DA930032.1 | - | 3 | 282 | |||

TM6SF2
Genomic Map