siRNA Details
| Gene Symbol: TM6SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001524.2 | NP_001001524.2 | 4, 5 | 456 | ||
| GenBank mRNA | AF255923.1 | - | 4, 5 | 405 | ||
| BC120986.1 | - | 4, 5 | 430 | |||
| BC120987.1 | - | 4, 5 | 430 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM6SF2| Gene Symbol: TM6SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001524.2 | NP_001001524.2 | 4, 5 | 456 | ||
| GenBank mRNA | AF255923.1 | - | 4, 5 | 405 | ||
| BC120986.1 | - | 4, 5 | 430 | |||
| BC120987.1 | - | 4, 5 | 430 | |||