siRNA Details
| Gene Symbol: C11orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300913.1 | NP_001287842.1 | 3, 4 | 523 | ||
| NM_022338.3 | NP_071733.1 | 3, 4 | 523 | |||
| XM_005274053.3 | XP_005274110.1 | 3, 4 | 698 | |||
| GenBank mRNA | AF264781.1 | - | 3, 4 | 483 | ||
| AY358754.1 | - | 1, 2 | 85 | |||
| BC011765.2 | - | 2, 3 | 372 | 3614861 | ||
| BG701401.1 | - | 3, 4 | 528 | |||
| BI862360.1 | - | 3, 4 | 497 | |||
| BX384793.2 | - | 2, 3 | 325 | |||

C11orf24
Genomic Map