siRNA Details
| Gene Symbol: RAB24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031677.3 | NP_001026847.1 | 6 | 906 | ||
| NM_130781.3 | NP_570137.2 | 7 | 820 | |||
| NR_109789.1 | 6 | 1038 | ||||
| NR_109790.1 | 5 | 1124 | ||||
| GenBank mRNA | AF087904.1 | - | 6 | 624 | ||
| AF258583.1 | - | 5 | 868 | |||
| AK021761.1 | - | 4 | 1155 | |||
| AK308155.1 | - | 6 | 993 | |||
| AL833898.1 | - | 6 | 448 | |||
| BC010006.2 | - | 7 | 743 | 4300377 | ||
| BC015534.1 | - | 7 | 782 | 3916832 | ||
| BC021263.2 | - | 6 | 866 | 4329216 | ||
| BT007268.1 | - | 6 | 450 | |||

RAB24
Genomic Map