siRNA Details
| Gene Symbol: CSNK1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022048.3 | NP_071331.2 | 9 | 1355 | ||
| GenBank mRNA | AB042562.1 | - | 8 | 1003 | ||
| AB042563.1 | - | 8 | 987 | |||
| AF223357.2 | - | 8 | 1073 | |||
| AK026957.1 | - | 5 | 462 | |||
| AK225960.1 | - | 9 | 1285 | |||
| AL832551.1 | - | 9 | 1133 | |||
| BC017236.2 | - | 8 | 804 | |||
| BC130613.1 | - | 9 | 1128 | 40146644 | ||
| BC136504.1 | - | 9 | 1128 | 9020493 | ||

CSNK1G1
Genomic Map