siRNA Details
| Gene Symbol: SLC37A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320537.1 | NP_001307466.1 | 6 | 1484 | ||
| NM_018964.3 | NP_061837.3 | 7 | 853 | |||
| XM_011529614.2 | XP_011527916.1 | 5 | 1625 | |||
| XM_011529615.1 | XP_011527917.1 | 5 | 1625 | |||
| XM_017028376.1 | XP_016883865.1 | 5 | 1625 | |||
| XM_017028377.1 | XP_016883866.1 | 7 | 778 | |||
| XM_017028378.1 | XP_016883867.1 | 7 | 1237 | |||
| XM_017028379.1 | XP_016883868.1 | 8 | 884 | |||
| XM_017028380.1 | XP_016883869.1 | 6 | 684 | |||
| XM_017028381.1 | XP_016883870.1 | 7 | 778 | |||
| XM_017028382.1 | XP_016883871.1 | 5 | 1966 | |||
| XM_017028383.1 | XP_016883872.1 | 7 | 709 | |||
| GenBank mRNA | AF311320.1 | - | 6 | 1423 | ||
| AJ269529.2 | - | 7 | 853 | |||

SLC37A1
Genomic Map