siRNA Details
| Gene Symbol: CLGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130675.1 | NP_001124147.1 | 11 | 1354 | ||
| NM_004362.2 | NP_004353.1 | 10 | 1270 | |||
| GenBank mRNA | AK093096.1 | - | 10 | 1833 | ||
| AK290819.1 | - | 10 | 1267 | |||
| AK299243.1 | - | 9 | 1059 | |||
| BC028357.2 | - | 10 | 1224 | 4830135 | ||
| D86322.1 | - | 10 | 1237 | |||

CLGN
Genomic Map