siRNA Details
| Gene Symbol: GPR88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022049.2 | NP_071332.2 | 2 | 2020 | ||
| GenBank mRNA | AB042410.1 | - | 2 | 2053 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR88| Gene Symbol: GPR88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022049.2 | NP_071332.2 | 2 | 2020 | ||
| GenBank mRNA | AB042410.1 | - | 2 | 2053 | ||
Set Membership: |
|