siRNA Details
| Gene Symbol: ERRFI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018948.3 | NP_061821.1 | 4 | 1224 | ||
| XM_005263477.2 | XP_005263534.1 | 4 | 1238 | |||
| XM_006710697.2 | XP_006710760.1 | 4 | 2408 | |||
| XM_011541596.2 | XP_011539898.1 | 3 | 1145 | |||
| GenBank mRNA | AJ276373.1 | - | 3 | 975 | ||
| AK027830.1 | - | 1 | 319 | |||
| AK096149.1 | - | 4 | 1199 | |||
| AK315718.1 | - | 4 | 1179 | |||
| AL137274.1 | - | 1 | 73 | |||
| BC025337.1 | - | 4 | 1187 | 4810371 | ||

ERRFI1
Genomic Map