siRNA Details
| Gene Symbol: TREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242589.2 | NP_001229518.1 | 3 | 657 | ||
| NM_018643.4 | NP_061113.1 | 3 | 657 | |||
| NR_136332.1 | 3 | 657 | ||||
| XM_011514696.1 | XP_011512998.1 | 3 | 597 | |||
| XM_017010956.1 | XP_016866445.1 | 3 | 597 | |||
| GenBank mRNA | AF196329.1 | - | 3 | 614 | ||
| AF287008.1 | - | 3 | 631 | |||
| AK130673.1 | - | 1 | 1075 | |||
| AK223264.1 | - | 3 | 594 | |||
| AK292162.1 | - | 3 | 594 | |||
| AK313104.1 | - | 3 | 609 | |||
| BC017773.1 | - | 3 | 612 | 4692680 | ||
| BI910026.1 | - | 3 | 577 | |||
| DA362458.1 | - | 2 | 104 | |||

TREM1
Genomic Map