siRNA Details
| Gene Symbol: FBXO42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018994.2 | NP_061867.1 | 2 | 416 | ||
| XM_006710698.3 | XP_006710761.1 | 1 | 189 | |||
| XM_017001502.1 | XP_016856991.1 | 2 | 379 | |||
| XM_017001503.1 | XP_016856992.1 | 2 | 309 | |||
| GenBank mRNA | AK055598.1 | - | 2 | 379 | ||
| AK310671.1 | - | 2 | 351 | |||
| AL833207.1 | - | 1 | 111 | |||
| BC043410.1 | - | 2 | 381 | 6155685 | ||
| BC063864.1 | - | 2 | 378 | 6182628 | ||
| DA488838.1 | - | 2 | 417 | |||
| HY007337.1 | - | 2 | 404 | |||

FBXO42
Genomic Map