siRNA Details
| Gene Symbol: TAF7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168474.1 | NP_001161946.1 | 10, 11 | 1026 | ||
| NM_024885.3 | NP_079161.3 | 10, 11 | 1194 | |||
| XM_005262152.1 | XP_005262209.1 | 10, 11 | 1029 | |||
| XM_006724664.1 | XP_006724727.1 | 10, 11 | 1196 | |||
| GenBank mRNA | AF285595.1 | - | 10, 11 | 1022 | ||
| AK308261.1 | - | 10, 11 | 1196 | |||
| BC043391.1 | - | 10, 11 | 1180 | 6154641 | ||

TAF7L
Genomic Map